Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

4-Carboxyphenylboronic acid ≥97%

Supplier: AOBChem USA

Warning
Synonyms: 4-Carboxybenzeneboronic acid4-Boronobenzoic Acid, Ark Pharm

10220-25G
77497-396EA 62.5 USD InStock
77497-396
4-Carboxyphenylboronic acid ≥97%
4-Carboxyphenylboronic acid
Caution: for Research Use Only


Formula: C₇H₇BO₄
MW: 165.94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00151801
CAS Number: 14047-29-1
Order Now


 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty