Would you like to log in again?
Supertek® Size of Particle Experiment
Supplier: Shiv Dial Sud & Sons
Supertek®
SOP1-C
470331-762EA 597.6 USD InStock
470331-762
Supertek® Size of Particle Experiment
Educational Materials Physics Educational Materials Optics Learning Activities
Measure the width/thickness of the given samples by analyzing the diffraction pattern.
  • Adjustable laser allows easy alignment of the laser source with the sample
  • A laser range finder allows for use of any wall as a screen, increasing the distance between the screen and the sample
  • Multiple samples can be tested and verified using modular setup
The characteristics of light such as interference and diffraction can be understood when light is studied as a wave phenomenon. Interaction of waves with matter results in either transmission, reflection, absorption, or diffraction of the wave. When the size of the matter is comparable to the wavelength of the wave that it interacts with, a phenomenon called diffraction occurs.
Diffraction of light due to particles is a function of the size of the particle and the wavelength of the light incident. It is possible to measure the size of a particle by studying the diffraction patterns created by it.
Delivery information: Kit includes optical bench set 0.8 m, laser source holder, grating holder, screen holder, laser distance meter, diffraction grating, frame with thread type 1, frame with thread type 2, frame with thread type 3.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty