Would you like to log in again?
Supertek® Faraday's Effect Experiment
Supplier: Shiv Dial Sud & Sons
Supertek®
FEE1-C
470331-750EA 1415.5 USD InStock
470331-750
Supertek® Faraday's Effect Experiment
Educational Materials Physics Educational Materials Optics Learning Activities
Observe the effect of a magnetic field on the polarization light in a dispersive medium.
  • Measure the Verdet’s constant of a given dispersive material
  • Modular U-Core design with precise optical alignment
  • Sensitive light meter allows precise measurement of the amount of rotation of plane polarization
When linearly polarized light passes through an optical medium in a region of the strong magnetic field, the plane of polarization of linearly polarized light rotates by an angle. The angle of rotation of plane-polarized light is proportional to the length of the optical medium and component of the magnetic field in the direction of light. The factor of proportionality is medium-specific and is called Verdet’s constant and this effect is known as the Faraday Rotation or Faraday Effect.
Discovered by Michael Faraday in 1845, the Faraday effect was the first experimental evidence that light and electromagnetism are related. In the experimental setup, the optical medium is an SF6 glass cube.
Delivery information: Kit includes optical bench set 0.8 m, light source holder, polarizer holder, analyzer holder, electromagnet set assembly, glass block, light sensor holder, Gauss meter, power supply for electromagnet, power supply for light source, data processor, DP switch.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty