Would you like to log in again?
Supertek® Malus' Law Experiment
Supplier: Shiv Dial Sud & Sons
Supertek®
MLE1-C
470331-758EA 727.25 USD InStock
470331-758
Supertek® Malus' Law Experiment
Educational Materials Physics Educational Materials Optics Learning Activities
Experimentally verify Malus' Law.
  • Optical bench makes precise optical alignment easy
  • Analyzer may be rotated precisely within 1° without disturbing the optical alignment
  • Sensitive digital lux meter enables precise measurement of light intensity (1 lux)
Light, when modeled as a wave phenomenon, can be classified as a transverse electromagnetic wave consisting of oscillating electric and magnetic fields that are oriented perpendicular to each other. Depending on the orientation of the plane of polarization of the electric field with respect to the direction of propagation of the wave, the wave can be classified as polarized or un-polarized. To measure the variation of transmission of an EM wave through two polarizers as a function of the angle of orientation between them and prove Malus’s Law.
Delivery information: Kit includes optical bench set 0.4 m, light source holder, polarizer holder, analyzer holder, light sensor holder, power supply for light source, data processor.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty