Would you like to log in again?
Supertek® Photoconductivity Experiment
Supplier: Shiv Dial Sud & Sons
Supertek®
PCE1-C
470331-760EA 534.05 USD InStock
470331-760
Supertek® Photoconductivity Experiment
Educational Materials Physics Educational Materials Electricity/Magnetism Learning Activities
Study the photoconductivity of CdS Photoresistor.
  • Optical bench makes precise optical alignment easy
  • Easy light intensity adjustment via two polarizers
  • Simple, color-coded electrical connections
The phenomenon of photoconductivity occurs when an incident light shown upon a semiconductor causes an increase in its electrical conductivity. This is because of the excitation of electrons across the energy gap into the conduction band, which leads to an increase in the number of free carriers in the conduction band, hence, an increase in the conductivity of the semiconductor. Here, we can study the characteristics of the CdS photoresistor under different conditions of light intensity and applied voltage.
Delivery information: Kit includes optical bench set 0.4 m, light source holder, polarizer holder, analyzer holder, LDR module holder, power supply, 0 to 15 V, 200 mA, power supply for light source, instruction manual.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty