Would you like to log in again?
Human Serum AB (Off The Clot), Gamma Irradiated
Supplier: Gemini Bio-Products
100-318G-100
77463-596EA 734.25 USD InStock
77463-596
Human Serum AB (Off The Clot), Gamma Irradiated
Sera (Serum)
Off the clot human serum AB is collected from normal healthy male donors at FDA-licensed facilities in the United States. As a natural off the clot product, this material contains no anticoagulants/preservatives. All donor units are tested for viral markers and found to be non-reactive.
  • Supports clinical and commercial applications
  • Aseptic assurance system
  • Gamma irradiated
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty