Would you like to log in again?
KIMBLE® KONTES® Flow Control Inlet Adapters with PTFE Plug, DWK Life Sciences
Supplier: DWK Life Sciences
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
KONTES®
185030-2942 275230-0000 275240-0000 185030-2440
KT275240-0000EA 208.25 USD InStock
KT275240-0000 KT185030-2942 KT275230-0000 KT185030-2440
KIMBLE® KONTES® Flow Control Inlet Adapters with PTFE Plug, DWK Life Sciences
Adapters Adapters, Joints with Tube Connectors
Designed to regulate the input of gases or liquids into reaction systems.
  • Stopcock with PTFE plug
  • Inner [ST] joint on one end
  • 90° hose connection on opposite end
The stopcock has an 821001-0002 PTFE plug
Manufactured from 33 expansion, low extractable borosilicate glass conforming to USP Type I and ASTM E438, Type I, Class A requirements.
Features an inner Standard Taper joint on one end and a 90° hose connection on the opposite end.
Order Now
ORDER
DOCUMENTATION
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty