Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
Cryogenic Sterile Microtubes, Bio Plas
Supplier: Bio Plas
4202S 4203S 4200S 4204S 4202S 4204S 4200S 4203S
20170-727PK 413.95 USD InStock
20170-727 20170-726 20170-703 20170-730 20170-722 20170-733 20170-732 20170-731
Cryogenic Sterile Microtubes, Bio Plas
Tubes Microcentrifuge Tubes
Designed for biological fluids stored in liquid nitrogen vapor.
  • Skirted models are freestanding and require the 20170-714 adapter
  • Tubes withstand over 20000×g
  • Gamma radiation sterilized
Ideal for specimens and cell cultures. Made of polypropylene copolymer, these tubes are leakproof, inert, non-pyrogenic, and virtually unbreakable. External threads eliminate material hang-up and minimize loss and contamination of valuable or hazardous samples. Screw cap has an internal O-ring for leakproof processing and storage. For use in microcentrifuges with fixed-angle heads.
Ordering information: Supplied sterilized and pre-capped.
Packaging: Packed 50/bag, 10 bags/pack, 2 packs/case (1000 each).
Order Now
ORDER
ACCESSORIES
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty