Would you like to log in again?
Lindberg/Blue M Single-Zone Tube Furnaces, 1500 °C, Thermo Scientific
Supplier: Thermo Scientific
Lindberg/Blue M
STF54453C STF55433C-1
52346-038EA 20983.09 USD InStock
30760-084 52346-038
Lindberg/Blue M Single-Zone Tube Furnaces, 1500 °C, Thermo Scientific
Ovens Tube Furnaces
Solid design utilizes double-end silicon carbide heating elements, positioned above and below the chamber on furnace 30760-084 and at right angles to the process tube on furnace 30777-006
Removable louvered panels provide easy access to heating elements and Type-R thermocouple. Chamber is constructed of high temperature grade refractory insulation. Insulating vestibules at each end of the chamber enhance overall temperature uniformity.
Ordering information: Furnace 30760-084 includes an integral, 16-segment programmable controller. Furnace 30777-006 requires an external controller, sold separately. Interconnecting power wires and process tubes are not included. An optional RS-485 communications data port is available on both models; contact your Avantor sales representative.
Order Now
ORDER
ACCESSORIES
SERVICES
Services: VWRCATALYST™ offers validation services including IQ, OQ, and PQ protocol development and execution, as well as asset management and calibration services (US only); call 888-793-2300 for more details.
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty