Would you like to log in again?
Please be aware that our e-commerce platform will be undergoing scheduled maintenance from 6:00 AM to 2:00 PM ET on May 11, 2024. During this time our site will be not available. We appreciate your patience.
National™ Autosampler Vial Racks, Thermo Scientific
Supplier: Thermo Scientific
C4012-25 C4011-25A
76212-314PK 87.27 USD InStock
76212-314 66030-198
National™ Autosampler Vial Racks, Thermo Scientific
Racks Chromatography Vial Racks
Stackable storage racks for 2 ml vials.
  • Polypropylene vial racks are resistant to most solvents
  • Efficiently hold and organize vials while using them or for storage
Racks feature alphanumeric indexing for easier vial identification. Save space by stacking the 50 or 100 plastic vial racks on top of each other.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty