Would you like to log in again?
Friedrichs Condenser, Chemglass
Supplier: Chemglass
CG-1210-01
80067-264EA 469.72 USD InStock
80067-264
Friedrichs Condenser, Chemglass
Condensers
Condenser has a specially formed spiral inner tube providing a long vapor path, ensuring good heat transfer and anti-flooding characteristics
Condenser has a [ST] outer joint at a 15° angle to the condenser and a lower inner drip tip joint. Hose connections have an O.D. of 10mm at the middle serration. [ST] joint: 24/40.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty