Would you like to log in again?
KleenGuard™ A10 Light Duty Coveralls, Kimberly-Clark Professional
Supplier: Kimberly-Clark
KleenGuard®
10636 12230 10621 10610
89132-912CS 88.88 USD InStock
89132-912 89132-926 89413-176 89132-918
KleenGuard™ A10 Light Duty Coveralls, Kimberly-Clark Professional
Disposable Garments
Light-duty, disposable coveralls provide protection against dirt, grime, and certain dry particulates in nonhazardous environments.
  • Light duty, disposable; protects against dirt, grime, and dry particulates in nonhazardous environments
  • KleenGuard™ A10 fabric is five layers of stacked barrier protection (SMMMS) against hazardous particles, offering economical protection
  • Lightweight fabric is breathable, cool, and comfortable
  • For use in food processing, manufacturing, and general maintenance
  • Coveralls adhere to ANSI/ISEA 101 sizing standards
This product is recyclable with the RightCycle® program by Kimberly-Clark Professional®.
Lightweight, spunbound polypropylene fabric is breathable, cool, and comfortable. Coveralls are best suited for use in food processing, manufacturing, and general maintenance applications. Available in three styles; zipper front closure with elastic wrists, zipper front closure with elastic wrists and attached hood, or zipper front closure with elastic wrists and attached hood and boots. Coveralls adhere to ANSI/ISEA 101-1996 sizing standards.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty