Would you like to log in again?
PREVAIL™ Flame-Resistant Coveralls, Kimberly-Clark®
Supplier: Kimberly-Clark
PREVAIL™
45322-00 45325 45316-00 45317-00 45352 45352-00 45315 45314-00 45323-00 45353 45312 45356-00 45313 45324 45315-00 45355-00 45314 45354 45323 45355 45354-00 45322 45327-00 45325-00 45316 45326-00 45324-00 45317 45312-00 45357 45357-00 45327 45326 45353-00 45356 45313-00
82006-140CS 452.14 USD InStock
82006-140 CA82006-130 82006-120 82006-142 82006-116 CA82006-124 CA82006-146 82006-138 82006-118 CA82006-126 CA82006-142 82006-134 CA82006-120 82006-112 82006-136 CA82006-122 CA82006-144 82006-114 CA82006-128 82006-130 CA82006-140 82006-132 CA82006-136 82006-126 CA82006-114 CA82006-138 CA82006-116 82006-128 82006-122 82006-144 CA82006-132 82006-124 CA82006-112 82006-146 CA82006-134 CA82006-118
PREVAIL™ Flame-Resistant Coveralls, Kimberly-Clark®
Disposable Garments
While not primary fire protection gear, these coveralls are designed to be worn in areas where exposure to sparks or flames is possible.
  • Comfortable, breathable, and durable fabric resists tearing
  • Design meets the ANSI/ISEA 101-1996 sizing standard
  • Blue fabric
Certifications: Coveralls are flame resistant compliant per NFPA 701 FLAME PROPAGATION TEST (1989 Edition), and in compliance with the flame resistance requirements cited by paragraph 6.1 of CAN/CGSB-155.20-2000. NFPA 99 Compliant Antistatic Material.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty