Would you like to log in again?
All-Aluminum Inner Seals, West Pharmaceutical Services
Supplier: West Pharmaceutical Services
5130-0066 5120-0321 5120020F 51206005
89008-984CS 1484.88 USD InStock
89008-984 89008-844 89012-310 89012-312
All-Aluminum Inner Seals, West Pharmaceutical Services
Closures Chromatography Vial Closures
For use with injection and infusion bottles, dental cartridges, X-ray imaging solutions, oral liquids, and opthalmic, veterinary, and agricultural applications
Plain aluminum ferrule seals feature lacquered shell and center-holes for crimping stoppers to glass vials. Seals have a turned-in-edge and are capable of high-speed processing.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty