Would you like to log in again?
Vacuum Filtration Adapters, Chemglass
Supplier: Chemglass
CG-1049-A-14 CG-1049-C-02 CG-1052-A-14 CG-1049-C-04 CG-1049-B-24 CG-1049-A-24 CG-1049-B-14 CG-1049-C-06 CG-1052-A-24
89049-146EA 223.07 USD InStock
89049-146 89049-144 89049-156 89049-148 89049-150 89049-142 89049-154 89049-140 89049-152
Vacuum Filtration Adapters, Chemglass
Adapters Adapters, Joints with Tube Connectors
Adapters are available in two bottom configuations and four top configurations.
  • Available bottoms include ground joint and threaded
  • Available tops include flare, threaded, and luer joint
  • Some models feature PTFE stopcock
Adapters with [ST] ground joint bottom connect to vials and flasks with [ST] outer joints
Adapters with outer luer joint top connect solid phase extraction (SPE) cartridges, tubes or syringes to a [ST] inner joint, allowing for easy sample concentration and simple clean-up, while reducing possibility of cross-contamination. Adapters 89049-144 and -146 have a 2mm PTFE stopcock between the outer and inner joints to control vacuum source. Adapters with heavy duty tooled flare top are for reduced pressure filtration with plain stem buchner funnels, and accept pluro stopppers (not included). Adapter 89049-156 has a 20-400 GPI threaded top for transfer from vial to vial.
Adapters with threaded bottoms have a 20-400 GPI thread for connection to vials (using a threaded connector) and a sealed-in drip tube to prevent breakage.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty