Would you like to log in again?
Condenser, Friedrichs, 250 mm, No. 3 Stopper Joint, Ace Glass Incorporated
Supplier: Ace Glass
5972-10
89053-890EA 413.85 USD InStock
89053-890
Condenser, Friedrichs, 250 mm, No. 3 Stopper Joint, Ace Glass Incorporated
Condensers
Friedrichs condenser with molded spiral condensing surface and standard taper inner joint at bottom
Top tubulation is tooled to take a No. 3 stopper. Jacket length is 250mm. Use with 3/8-inch I.D. tubing, size D hose connections.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty