Would you like to log in again?
10-425 Screw-Thread Vial Kits, SUN-SRi
Supplier: SUN SRi
REVIEWS (0)
Total Ratings: 0
Avg. Ratings: 0.0 out of 5
Sort all by:
200 438 200 403
66008-254PK 172.22 USD InStock
66008-254 14234-916
10-425 Screw-Thread Vial Kits, SUN-SRi
Vials
Kits are an economical alternative to ordering multiple components.
Preassembled vial kits feature vials, caps, and septa that are completely assembled to reduce the possibility of contaminating the sample vial. Convenient for special applications in which samples may be injected through the septum.
Unassembled vial kits feature preassembled caps and septa to save time during sample preparation and to minimize contamination during assembly. Vials are conveniently provided in an adjacent well for easy access.
Kits feature clear glass vials and black polypropylene screw caps with PTFE/silicone septa. Usable volume: 1.5 ml.
Order Now
ORDER
WRITE A REVIEW
REVIEWS (0)
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty