Would you like to log in again?
Infekta-Pak 1 Infectious Specimen Shippers, InfeKta
Supplier: InfeKta
Infekta-Pak Infekta-Pak 1
INF-1-4 INF-1
89030-506CS 297.01 USD InStock
89030-506 89030-504
Infekta-Pak 1 Infectious Specimen Shippers, InfeKta
Mailers and Shippers
Cost-efficient combination packages are designed to ship ambient infectous medical specimens of all risk groups
Shipper 89030-504 components include a 10.8Wx10.8Dx18.4Lcm (41/4x41/4x71/4") preprinted outer shipping box, a 7.9dia.x16.2Hcm (31/8x63/8") white fiberboard mailing tube with a seamed metal bottom and metal screw-cap lid, and a 95kPa, 6.4dia.x15.9Lcm (21/2x61/4") HDPE plastic bottle with a lined plastic screw cap. Outer box design also features a "B" flute perimeter insert to hold the mailing tube securely in place during transport.
Four-pack shipper 89030-506 components include a 21.6Wx21.6Dx18.4Hcm (81/2x81/2x71/4") preprinted outer shipping box with a four-chamber corrugated divider insert to separate each canister set, four 7.9dia.x16.2Lcm (31/8x63/8") white fiberboard mailing tubes with seamed metal bottoms and metal screw-cap lids, and four 95kPa, 6.4dia.x15.9Lcm (21/2x61/4") HDPE plastic bottles with matching lined plastic screw caps.
Five-hole foam insert 89030-538 is constructed of die-cut hydrophilic polyurethane foam and includes a 1.9cm (3/4") protective cover. It is designed to fit in shippers 89030-504 and -506 and can accommodate the most commonly used sizes of blood collection tubes.
Outer box is preprinted with UN certification registration, Class 6.2 infectious substance label, "to" and "from" address, and direction arrow to reduce the risk of shipping error due to improper labeling.
Ordering information: Optional foam inserts and absorbents are available separately. A shipper's declaration form is required (not included) and must accompany the shipment of these packages.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty