Would you like to log in again?
EP Preserved™ Vials with Preservatives, Thermo Scientific
Supplier: Thermo Scientific
EP Preserved™
PP140-40CEPPTTW
89039-948CS 771.12 USD InStock
89039-948
EP Preserved™ Vials with Preservatives, Thermo Scientific
Vials
Level 1 glass vials for use in testing for Volatile Organic Compounds (VOC).
Vials are tare-weighed and have pre-applied labels. Propylene caps are open-top with 0.125 PTFE/silicone septa. Capacity: 40 ml. Height: 9.5 cm (3³/₄"). Ø: 2.9 cm (1¹/₈"). Cap Size: 24-414.
Certifications: Vials comply with EPA SW846 Method 5035 requirements.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty