Would you like to log in again?
ACIDULATE 28 Cleaning Compound, Quip Laboratories
Supplier: Quip Laboratories
ACIDULATE 28
AD028-55GAL AD028-30GAL AD028-5GAL AD028-15GAL
21840-000EA 459 USD InStock
21840-000 21840-002 21840-004 21840-006
ACIDULATE 28 Cleaning Compound, Quip Laboratories
Cleaning Agents Detergents and Surface Cleansers
Designed to penetrate and dissolve urine and mineral scale deposits, and remove animal fat and protein soils.
Chemically-balanced cleaning compound blends citric acids, special wetting agents and detergents. Can be used in dip tanks or washing equipment. Use as a primary wash agent or as a secondary application. A 1% solution has a pH of 2.1. Compound is free rinsing and will not leave streak-forming residues. Biodegradable.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty