Would you like to log in again?
Vacuum Trap, Solvent Vapor, Plastic-Coated, Chemglass
Supplier: Chemglass
CG-4519-01
80063-072EA 1791.95 USD InStock
80063-072
Vacuum Trap, Solvent Vapor, Plastic-Coated, Chemglass
Vacuum Traps
Trap is designed for large volume solvent vapor removal during rotary evaporation.
Trap is placed between the vacuum source and the rotary evaporator to prevent solvent vapors from entering the vacuum pump. Plastic-coated to contain solvent in case of breakage and supplied complete with a UHMW lid with handle.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty