Would you like to log in again?
Diagna-Pak Diagnostic/Biological Substance–Category B Shippers, InfeKta
Supplier: InfeKta
Diagna-Pak
INFDIAG-20B INFDIAG-17B
89030-514CS 650.11 USD InStock
89030-514 89030-516
Diagna-Pak Diagnostic/Biological Substance–Category B Shippers, InfeKta
Mailers and Shippers
Combination packages consist of a white outer fiberboard screw-cap mailing tube with an inner HDPE plastic bottle.
The outer mailing tube features a seamed metal bottom, crimped metal neck, and matching threaded metal screw cap. The 95kpa inner bottle is supplied with a lined plastic white screw cap. Shipper 89030-514 will accommodate a 50mL conical centrifuge tube as a primary container (not included), and optional BioSorb® absorbent strips (see the 89030-530 series). Shipper 89030-516 will accommodate five-hole protective foam insert 89030-538 (sold separately). Insert is constructed of die-cut hydrophilic polyurethane foam and includes a 1.9cm (3/4") protective cover. It can accommodate the most commonly used sizes of blood collection tubes.
Certifications: Shippers meet PI 650 requirements.
Ordering information: A UN 3373 Diagnostic/Biological Substance–Category B specimen label (sold separately) is required with use of these shippers. Contact your Avantor sales representative for more information.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty