Would you like to log in again?
Aerostat™ PC Ionizing Air Blower, Simco Ion
Supplier: Simco
Aerostat®
4003368 4003367
56604-004EA 2075.91 USD InStock
56604-004 14214-052
Aerostat™ PC Ionizing Air Blower, Simco Ion
Ionizers
Compact, reliable ion air blower provides uncompromising fast static discharge over a work surface
No calibration is necessary; the unit’s balance circuitry maintains an inherent balance of ±5V. A built-in emitter point cleaner simplifies maintenance. An integrated heater provides user comfort, and fan speed control provides infinitely adjustable airflow from 14–28L/s (35–70cfm).
Certifications: The 120V model is UL and C-UL listed. The 230V model is CE marked.
Order Now
ORDER
SPECIFICATIONS
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty