Would you like to log in again?
Foaming Glass Cleaner, Techspray™
Supplier: ITW Chemtronics
Techspray™
1625-18S
14216-040CS 155.38 USD InStock
14216-040
Foaming Glass Cleaner, Techspray™
Cleaning Agents Detergents and Surface Cleansers
Fast-drying, non-streaking glass cleaner removes hand oils, ink marks, dust, fingerprints, and more from CRTs, computers, and other hard surfaces
Protects against static build-up.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty