Would you like to log in again?
Spor-Klenz® Concentrated Sterilant/Disinfectant, STERIS®
Supplier: Steris
Spor-Klenz®
652026
21899-861CS 599.36 USD InStock
21899-861
Spor-Klenz® Concentrated Sterilant/Disinfectant, STERIS®
Cleaning Agents Disinfectants
This fast-acting, liquid cold sterilant/disinfectant is a stabilized blend of peracetic acid, hydrogen peroxide, and acetic acid for use wherever critically high-level microbial control is required.
Filtered to 0.22µm, it is specifically formulated for use in sterilization and disinfection of hard surfaces. Low toxicity profile adds to worker safety. The product is sporicidal in 11 hours and disinfects in 10 minutes. Each 946mL (1qt.) bottle of concentrate makes 94.6L (25gal.) of solution at a concentration of 1:99. Certificates of analysis, filtration, and sterility are electronically available for each lot.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty