Would you like to log in again?
Combination Soil Outfit, LaMotte
Supplier: LaMotte
5010-01
89004-842EA 1446.62 USD InStock
89004-842
Combination Soil Outfit, LaMotte
Environmental Test Kits
This advanced reagent system and unique extraction procedure based on the Mehlich I extraction provides fast, simple, and accurate soil testing
Colorimetric test methods are used for most test factors. Tests for calcium, sulfate, and chlorides are based on turbidity measurements. Potassium analysis also employs a turbidity measurement, which uses a unique reading device designed to read directly in pounds per acre. A single extraction procedure, using Morgan universal extraction solution, provides the liquid soil extract for all of the nutrient tests with the exception of chloride, which is extracted with demineralized water. The humus screening test, performed on a soil sample-demineralized water suspension, uses five color standards for rapid measurement of humus content. Soil pH is determined colorimetrically using a series of pH indicators and color charts. Also included are simplified procedures for screening nitrates, phosphorus, and potassium in plant tissues.
Outfit contains 50 test of each type except pH which has 100.
A series of rapid, accurate chemical tests use standardized reagents to produce color reactions measured against laminated color charts.
Ordering information: Kit is furnished with a lightweight carrying case with all components securely mounted in removable foam trays. Each kit includes complete instructions, a pad of soil analysis report forms, and a soil management handbook containing information on interpreting test results for determining lime and fertilizer requirements. For complete reagent refill packages, contact your Avantor sales representative for more information.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty