Would you like to log in again?
QB2HYG® Banded Earplugs, Honeywell Safety
Supplier: Honeywell Safety
QB2HYG®
QB2HYG QB2HYG
46000-096CS 1080.23 USD InStock
CA46000-096 46000-096
QB2HYG® Banded Earplugs, Honeywell Safety
Earplugs Banded Earplugs
Quiet band hearing protector is a comfortable alternative to earplugs or muffs.
  • Offers highest attenuation among supra-aural hearing protectors
  • Lightweight, portable, and perfect when noise hazards are intermittent
  • Soft pods rest partially in the ear without excessive pressure on the ear canal
  • Design prevents ear pods from touching dirty or contaminated surfaces
  • Reusable when cleaned with soap and water
Noise reduction rating: 25dB
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty