Would you like to log in again?
Iron VACUettes® Test Kit, CHEMetrics®
Supplier: CHEMetrics
VACUettes®
R-6001B K-6010B
CXR-6001BPK 62.48 USD InStock
CXR-6001B CXK-6010B
Iron VACUettes® Test Kit, CHEMetrics®
Environmental Test Kits
Ampules are self-filling and designed for highly concentrated samples
The iron test tests for total and soluble iron using phenanthroline. Sensitivity for the total iron test is 0–120 ppm. Sensitivity for the soluble iron test is 120–1200 ppm.
The auto-dilution feature eliminates the need for time-consuming and error-prone preliminary dilutions. Each ampule has a capillary pipet attached to the tip that is calibrated to draw the correct volume for one of four dilution factors: 25X, 50X, 100X, and 1000X. Once the sample has been drawn, snap the tip, and results will appear in 2–3 minutes. Results can be quantified with a color comparator and provide an accuracy equal to a volumetric procedure that requires much more time.
Ordering information: The kit comes packaged in a plastic carrying case. The case contains 30 auto-dilution ampules, 5mL sample cup and lid, dilutor snapper cup, low and high range comparators, activator solution, micro test tube, instructions, and MSDS.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty