Would you like to log in again?
CrushTube™ Swab, Texwipe®
Supplier: Texwipe
CrushTube™
TX726
TWTX726CS 967.69 USD InStock
TWTX726
CrushTube™ Swab, Texwipe®
Swabs Swabs for Cleaning
Ideal for cleaning ionizer tips
Point-of-use IPA-saturated polyester swab has a polyester brush tip with a breakable glass ampule containing 91% isopropyl alcohol (IPA). When ampule is crushed using protective plastic sleeve, the brush tip becomes saturated and is ready for use. Silicone-free.
Ordering information: Packaged 50 swabs per bag, 10 boxes per case for a total of 500 swabs per case. Bag-Within-A-Bag® packaging.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty