Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Copper sulfate 0.100 M (M/10)

Supplier: Ricca Chemical
Synonyms: Cupric sulphateCopper(II) sulfateCopper(II) sulphateCupric sulfate

R2351000-4A
RCR2351000-4AEA 318.39 USD InStock
RCR2351000-4A
Copper sulfate 0.100 M (M/10)
Copper(II) sulphate solutions


Formula: CuSO₄·5H₂O
MW: 249.69 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00149681
CAS Number: 7758-99-8
Order Now
Specification Test Results
Appearance Blue liquid
Assay (vs. Sodium Thiosulfate/Starch) 0.0999-0.1001 Molar

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty