Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Sodium tetraphenylborate 0.02 M

Supplier: Ricca Chemical
Synonyms: Sodium tetraphenylboronTetraphenyl sodium borateTetraphenylboron sodium salt

R7855000-1A
RCR7855000-1AEA 127.97 USD InStock
RCR7855000-1A
Sodium tetraphenylborate 0.02 M
Sodium tetraphenylborate


Formula: (C₆H₅)₄BNa
MW: 342.22 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011494
CAS Number: 143-66-8
Order Now
Specification Test Results
Appearance Colorless to tan liquid
Assay (Calculated) 0.0198-0.0202 N at 20°C

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty