Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Toluidine blue 1% (w/v) stain

Supplier: Ricca Chemical

R8654000-1C
RCR8654000-1CEA 198.57 USD InStock
RCR8654000-1C
Toluidine blue 1% (w/v) stain
Toluidine blue


Formula: C₁₅H₁₆ClN₃S·0.5ZnCl₂
MW: 373.97 g/mol
Storage Temperature: Ambient
MDL Number: MFCD03548293
CAS Number: 6586-04-5
Order Now
Specification Test Results
Appearance Bluish-purple liquid
UV/Visible Spectrum To Pass Test

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty