Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Zinc sulfate 0.0200 M (M/50)

Supplier: Ricca Chemical

R9657000-500A
RCR9657000500AEA 33.06 USD InStock
RCR9657000500A
Zinc sulfate 0.0200 M (M/50)
Zinc sulphate


Formula: O₄SZn
MW: 161.45 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00011302
CAS Number: 7733-02-0
Order Now
Specification Test Results
Appearance Colorless liquid
Assay (vs. EDTA/Xylenol Orange) 0.01998-0.02002 Molar

 

ORDER

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty