Would you like to log in again?
Accessories for Synthware Repurification Solvent Still, Kemtech America
Supplier: Kemtech America
S211444
60005-142EA 44.57 USD InStock
60005-142
Accessories for Synthware Repurification Solvent Still, Kemtech America
Distillation Components Distillation Heads
Kemtech replacement t-bore stopcock.
  • Borosilicate 3.3
  • Double oblique
  • 1:5 taper
Double oblique, Teflon®, stopcock is a direct replacement for a number of Kemtech products.
Order Now
ORDER
ACCESSORIES
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty