Would you like to log in again?
Fectoggles™ Series Safety Goggles, 3M™ Company
Supplier: 3M
Fectoggles™
7010045739 7010045743
55666-245PK 362.73 USD InStock
55666-245 55666-252
Fectoggles™ Series Safety Goggles, 3M™ Company
Goggles
The reliable combination of safety goggles and glasses, Fectoggles™ are an innovation in protective eyewear.
  • Aspheric lens helps provide expansive coverage and an extended viewing range
  • Foam flange seals the perimeter of the lens to help keep out particulates and sweat
  • Fire-retardant, braided, elastic strap holds the goggles firmly in place
A foam flange seals the perimeter of the lens to help keep out particulates and sweat, while a fire-retardant, braided, elastic strap holds the goggles firmly to the wearer's profile. This unique lens has no parting lines to obstruct view and is adaptable for indoor or outdoor use. These goggles do not provide splash protection.
Certifications: Meets ANSI Z87.1-2010, CSA Z94.3-2007.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty