Would you like to log in again?
SP Bel-Art Desiccant in a Cartridge, Bel-Art Products, a part of SP
Supplier: Bel-Art
F42045-0200 F42045-0160
24988-324EA 139.77 USD InStock
24988-324 24988-302
SP Bel-Art Desiccant in a Cartridge, Bel-Art Products, a part of SP
Desiccators Desiccants
Convenient, easy-to-handle cartridges with Natrasorb® TI indicating silica gel beads fit Scienceware® “Space Saver” desiccators, in addition to other plastic desiccators of similar size.
  • Easy to handle
  • Beads change color with moisture content
  • Durable polyester felt bag
Beads change color from blue when dry to pink when moist and are contained in a durable, polyester felt bag with a perforated Mylar® cover sealed in place.
Saturated gel can be regenerated in a 150ºC (300ºF) vented oven for 3 hours or more (depending on size) until blue color returns. May be regenerated repeatedly without degradation.
Ordering information: Individually sealed in moisture proof pouches.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty