Would you like to log in again?
OX™ Protective Eyewear, 3M™
Supplier: 3M
OX™
7000002373
47744-993CS 205.72 USD InStock
47744-993
OX™ Protective Eyewear, 3M™
Safety Glasses
Sturdy, durable OX™ Protective Eyewear delivers a flexible, polycarbonate construction with over-the-glass coverage.
  • Lightweight and adjustable over-the-glasses
  • SecureGrip™ temples to fit virtually any size face
  • Extends the field of vision due to a wide profile
  • Seamless lens features built-in sideshields to secure an uninterrupted sight range with Over-the-glass coverage
OX™ Protective Eyewear increases comfort and provides custom fit and adjustability for prescription eyeglass wearers.
Certifications: ANSI Z87.1-2003. / CSA Z94.3-2007
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty