Would you like to log in again?
Uvex Astrospec 3000® Welding Eyewear, Honeywell Safety
Supplier: Honeywell Safety
Astrospec 3000®
S1111 S1111
14231-226EA 0.22 USD InStock
CA14231-226 14231-226
Uvex Astrospec 3000® Welding Eyewear, Honeywell Safety
Safety Glasses
Highly adjustable protective eyewear with a face-hugging fit and panoramic field of vision.
  • Lightweight, impact-resistant polycarbonate unilens
  • Adjustable lens angle for maximum comfort
  • Lenses are easily and economically replaceable
  • Molded-in sideshields and brow guard enhance protection without obstructing peripheral vision
  • Clear lens with a thickness of 2.3 mm offers 385 nm UV protection for indoor work.
Nylon Duoflex® frame with four-stop temple adjustment (130 to 150 mm) is designed so that temples will not scratch lens when eyewear is folded.
The Ultra-dura® lens coating provides superior scratch and chemical resistance. The Uvextreme® AF lens coating provides antifog, anti-scratch, antistatic, and anti-UV protection.
Certifications: ANSI Z87.1-1989 / CSA Z94.3-1992
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty