Would you like to log in again?
Uvex Millennia™ Protective Eyewear, Honeywell Safety
Supplier: Honeywell Safety
uvex millennia™
11150350 11150354 11150370 11150353 11150350
32999-818EA 13.94 USD InStock
32999-818 32999-809 32999-806 32999-800 CA32999-800
Uvex Millennia™ Protective Eyewear, Honeywell Safety
Safety Glasses
Uvex Millennia™ safety eyewear combines futuristic wrap-around styling with exceptional coverage and protection. Comfortable and lightweight, the face-contouring frame provides exceptional brow and side protection for the most demanding work environments.
  • Wrap-around unilens design, comfortable universal nosebridge
  • High impact dual 9-base wrap-around unilens
  • Clear lenses
Available with an extensive selection of high-performance lens tints and coatings, the Uvex Millennia™ is a popular choice among workers.
One-piece lens features an anti-scratch coating. Glasses include a comfortable universal nosepiece and an adjustable break-away Flexi-Cord™ neck cord for added security and convenience.
Certifications: Meets ANSI Z87+ (High Impact) and tested by Honeywell to meet the requirements of the CSA Z94.3 standard. Meets military V0 ballistic test for impact.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty