Would you like to log in again?
Uvex Falcon® Protective Eyewear, Honeywell Safety
Supplier: Honeywell Safety
uvex falcon®
S4500X S4500X
14222-884CS 2787.74 USD InStock
14222-884 CA14222-884
Uvex Falcon® Protective Eyewear, Honeywell Safety
Safety Glasses
Eyewear features a dual nine-base wrap-around unilens for superior peripheral coverage and protection of the entire eye area.
  • Lightweight frame
  • Adjustable nylon temples
  • Clear lens with Uvextreme® lens coating for antifog, anti-scratch, antistatic, and anti-UV protection
Glasses provide workers with a clear and unobstructed view of the work environment.
Certifications: ANSI Z87+ /CSA Z94.3
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty