Would you like to log in again?
SMITH & WESSON® 38 SPECIAL™ Safety Eyewear, KIMBERLY-CLARK PROFESSIONAL®
Supplier: Kimberly-Clark
Smith & Wesson®
19855 19857 19859
67200-216EA 6.02 USD InStock
67200-216 67200-218 67200-212
SMITH & WESSON® 38 SPECIAL™ Safety Eyewear, KIMBERLY-CLARK PROFESSIONAL®
Safety Glasses
Bold and stylish safety glasses feature a precision frame with extended temples.
  • Ergonomically contoured temples with grip rubber inserts for a secure and comfortable fit
  • Shatter-resistant polycarbonate lenses
  • 99.9% UV protection
Available in a variety of frame colors and lens tints.
Certifications: ANSI Z87.1+
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty