Would you like to log in again?
Personal Diffusional Air Sampling Monitors, 3M™
Supplier: 3M
7000126238 7000126239 7000051778
56614-641CS 8606.14 USD InStock
56614-641 67100-734 67100-736
Personal Diffusional Air Sampling Monitors, 3M™
Gas Detection Systems
Monitors use the proven scientific principle of diffusion, which is the gradual mixing of molecules of two or more substances.
Ethylene Oxide Monitor utilizes brominated carbon to collect and convert the reactive ethylene oxide to a more stable chemical derivative, 2-bromoethanol. This compound is then desorbed with methylene chloride in methanol. It meets OSHA's requirement for accuracy within ±25% for 8 hour samplings at 1ppm and the requirement for accuracy within ±35% for 8 hour samplings at the action limit.
Formaldehyde Monitor uses a bisulfite-impregnated filter and is analyzed by colorimetry. It can be used for 8 hour TWA sampling to meet the OSHA requirement of ±25% accuracy at 0.5–5.0ppm. However, this monitor does not have the sensitivity for short term exposure limit (STEL) monitoring. Monitor with included pre-paid analysis and writen report option is available in the US market.
Caution: All 3M™ products must be used in accordance with OSHA regulations and the user instructions, warnings, and limitations accompanying each product.
They are designed for sampling personal exposures in work areas in a variety of industries.
Order Now
ORDER
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty