Would you like to log in again?
Calbiochem® Bilirubinm
Supplier: EMD Millipore
Calbiochem®
2011-1GM 2011-1GM
80057-868EA 370.63 USD InStock
CA80057-868 80057-868
Calbiochem® Bilirubinm
Proteins and Peptides
Principal pigment of bile and one of the major end products of hemoglobin decomposition. Contains conjugated double bonds and reactive hydrogen atoms. Proposed to serve as a physiological antioxidant. Reported to pass through the blood-brain barrier and exert significant antioxidant effects in the brain. Extinction coefficient (454nm, trichloromethane): 59,100–62,300 M-1cm-1.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty