Would you like to log in again?
Calbiochem® Human Ferritin
Supplier: EMD Millipore
Calbiochem®
341482-1MG 341482-1MG
80056-442EA 872.32 USD InStock
80056-442 CA80056-442
Calbiochem® Human Ferritin
Proteins and Peptides
Sterile-filtered. A major iron storage protein. Suitable for use in immunoassays, as an immunogen, and in enzyme/radiolabeling. Prepared from tissues of individuals that have been shown by certified tests to be negative for HBsAg and for antibodies to HIV and HCV.
Order Now
ORDER
SPECIFICATIONS
DOCUMENTATION
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty