Would you like to log in again?
Calbiochem® Human Recombinant Interleukin 1A (from E. coli)
Supplier: EMD Millipore
Calbiochem®
407611-2UG
80052-584EA 494.54 USD InStock
80052-584
Calbiochem® Human Recombinant Interleukin 1A (from E. coli)
Proteins and Peptides
Potent stimulator of bone resorption and stimulator of nuclear phospholipase C. Induces nitric oxide synthase. Biological Activity: ED50=3–10pg/mL as measured in a cell proliferation assay.
Order Now
ORDER
SPECIFICATIONS
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty