Would you like to log in again?
Crews® Hombre™ Protective Eyewear, MCR Safety
Supplier: MCR Safety
Crews®
HB119 HB110AF HB112 HB110 HB112AF
80081-883EA 15.77 USD InStock
80081-883 80081-891 80081-881 80081-887 80081-889
Crews® Hombre™ Protective Eyewear, MCR Safety
Safety Glasses
These stylish glasses feature a dual-focus, polycarbonate single-lens design ensuring nearly complete orbital eye protection.
Polycarbonate lenses with Duramass® scratch-resistant coating filter out 99% of UV radiation. Antifog lenses feature Duramass AF4® antifog protection. Durable, matte finished frame consists of a bayonet temple design and features nonslip rubber head grips. Soft, secure gel nosepiece provides a comfortable fit.
Certifications: Meet or exceed ANSI Z87.1 safety standards.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty