Would you like to log in again?
Fuse™ Protective Eyewear, Honeywell Safety
Supplier: Honeywell Safety
Fuse™
11150801 11150355 11150800 11150800 11150805
89030-543EA 30.74 USD InStock
89030-543 16003-872 16003-866 16003-868 CA16003-866
Fuse™ Protective Eyewear, Honeywell Safety
Safety Glasses
A 9-base wraparound design provides a distortion-free 180° field of vision. The frame features a nickel-free topcoat. Lenses are made from optical-grade polycarbonate with a protective hardcoat to maximize the life of the eyewear. Glasses are available with Fog-Ban® antifog coating, and include an adjustable breakaway FlexiCord™ neck cord for added security and convenience.
  • Soft, adjustable saddle nose pads
  • Flexible temple tips for comfortable fit
  • Lightweight and durable
Certifications: Meets ANSI Z87+ and CSA Z94.3 standards.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty