Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Dihydrolipoic acid ≥95% (by TLC), Calbiochem®, Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: DHLA6,8-Dimercapto-n-octanoic acidDL-α-Lipoic Acid, Dihydro

M-Clarity™ quality level = MQ100

437694-50MG 437694-50MG
80056-972EA 101.5 USD InStock
CA80056-972 80056-972
Dihydrolipoic acid ≥95% (by TLC), Calbiochem®, Sigma-Aldrich®
Dihydrolipoic acid
An effective antioxidant

Scavenges hypochlorous acid, peroxyl, superoxide, hydroxyl and nitric oxide radicals. Also exhibits pro-oxidant properties, as demonstrated by its ability to reduce iron ions and to generate reactive sulfur-containing radicals that can damage certain proteins, such as [alpha]1-antiproteinase and creatine kinase. DHLA has recently been shown to inhibit apoptosis in rat thymocytes induced by methylprednisolone and etoposide.


Formula: C₈H₁₆O₂S₂
MW: 208.33 g/mol
Storage Temperature: Refrigerator
MDL Number: MFCD00166974
CAS Number: 462-20-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty