Keep your session open?
Ending In 
Your shopping session has expired.
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

(S)-(+)-Camptothecin (from Camptotheca acuminata), Sigma-Aldrich®

Supplier: MilliporeSigma
Synonyms: 4-Ethyl-4-hydroxy-1H-pyrano-[3',4':6,7]indolizino[1,2-b]quinoline-3,14(4H,12H)-dione(+)-Camptothecin

208925-50MG 208925-50MG
80055-998EA 145.02 USD InStock
CA80055-998 80055-998
(S)-(+)-Camptothecin (from Camptotheca acuminata), Sigma-Aldrich®
(S)-(+)-Camptothecin
A DNA topoisomerase I inhibitor

Possesses antileukemic and antitumor properties. Induces apoptosis of HL-60 cells and mouse thymocytes.


Formula: C₂₀H₁₆N₂O₄
MW: 348.36 g/mol
MDL Number: MFCD00081076
CAS Number: 7689-03-4
Order Now

 

ORDER

DOCUMENTATION

Q and A

Product Questions and Answers

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty