Would you like to log in again?
Uvex Genesis X2™ Safety Eyewear, Honeywell Safety
Supplier: Honeywell Safety
uvex genesis X2™
S3523 S3522 S3500 S3500X S3532X S3524 S3504 S3512X S3521 S3500X S3521X
94023-952EA 26.59 USD InStock
94023-952 94023-950 94023-922 94023-944 94023-932 94023-920 94023-942 94023-938 94023-948 94023-946 CA94023-922
Uvex Genesis X2™ Safety Eyewear, Honeywell Safety
Safety Glasses
Exceptional comfort, protection, and modern style characterize the sport-inspired design of the Genesis X2 line of safety glasses
  • Gray lenses decrease eye fatigue outdoors
  • A dual 9-base wraparound lens provides excellent peripheral vision and side protection
  • Ratcheting temples permit users to customize lens inclination
Lenses are available in a variety of task-specific tints with either scratch- and chemical-resistant Ultra-dura® coating, or anti-fog Uvextreme® coating. Amber lenses enhance contrast in low-light environments. Espresso and gold mirror lenses minimize strong sunlight and glare. SCT-Blue lenses help counteract sodium vapor lighting. SCT-Reflect 50 lenses have a light mirror coating for varying light conditions. SCT-Vermillion lenses provide exceptional contrast indoors and are well-suited for inspections.
Soft, flexible fingers adjust and conform to virtually any nasal profile to reduce slippage, while an integrated elastomer brow guard works to diffuse and deflect impact.
Certifications: ANSI Z87+ standard and Mil V0 ballistic impact test.
Delivery information: Supplied with a lifetime frame guarantee.
Order Now
ORDER
Q and A
Product Questions and Answers
Enter Certificate Details
Enter the Lot number:
Choose from recent batch/lot numbers:
Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your cart, enter a quantity and click Add to Cart.
Original Product:
Description Catalog Number Availability Unit Your Price Price Per Qty